Naphthol Yellow S Hydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Flavianic Acid Disodium Salt8-Hydroxy-5,7-dinitro-2-naphthalenesulfonic Acid Disodium SaltDisodium 8-Hydroxy-5,7-dinitro-2-naphthalenesulfonate2,4-Dinitro-1-naphthol-7-sulphonic Acid Disodium SaltDisodium 2,4-Dinitro-1-naphthol-7-sulfonate2,4-Dinitro-1-naphthol-7-sulfonic Acid Disodium Salt

F0100-25G
TCF0100-025GEA 115 USD
TCF0100-025G
Naphthol Yellow S Hydrate ≥98.0% (by HPLC, titration analysis)
Naphthol yellow S hydrate

CAS Number: 846-70-8
MDL Number: MFCD00003958
Molecular Formula: C10H6N2O8S
Molecular Weight: 358.19
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Lambda max.: 429 nm (H2O)

Formula: C₁₀H₆N₂O₈S·xH₂O
MW: 358.19 g/mol (anh)
MDL Number: MFCD00003958
CAS Number: 846-70-8
https://us.vwr.com/stibo/web/std.lang.all/87/28/28968728.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR