Rose Bengal
Supplier: TCI America
Synonyms:
Rose Bengal
F0001-25G
TCF0001-025GEA
146.16
USD
TCF0001-025G
Rose Bengal
Rose Bengal
CAS Number: 632-69-9
MDL Number: MFCD00151169
Molecular Weight: 1017.63
Form: Powder
Color: Red
Formula:
C₂₀H₄Cl₄I₄O₅ MW: 1017.63 g/mol |
MDL Number:
MFCD00151169 CAS Number: 632-69-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...