Ethoxytriethylsilane ≥98.0%

Supplier: TCI America

E0790-25ML
TCE0790-25MLEA 142.1 USD
TCE0790-25ML
Ethoxytriethylsilane ≥98.0%
Ethoxytriethylsilane

CAS Number: 597-67-1
MDL Number: MFCD00054718
Molecular Formula: C8H20OSi
Molecular Weight: 160.33
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Specific Gravity (20/20): 0.81

Formula: C₈H₂₀OSi
Boiling Pt: 155 °C
Density: 0.81 (20 °C)
MDL Number: MFCD00054718
CAS Number: 597-67-1
https://us.vwr.com/stibo/web/std.lang.all/81/91/28968191.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR