Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyltrimethoxysilane ≥97.0%

Supplier: TCI America


E0789-25G
TCE0789-25GEA 28.04 USD InStock
TCE0789-25G
Ethyltrimethoxysilane ≥97.0%
Ethyltrimethoxysilane
CAS Number: 5314-55-6
MDL Number: MFCD00051541
Molecular Formula: C5H14O3Si
Molecular Weight: 150.25
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 124
Flash Point (°C): 27
Specific Gravity (20/20): 0.95


Formula: C₅H₁₄O₃Si
MW: 150.25 g/mol
Boiling Pt: 124 °C
Density: 0,95 (20 °C)
Flash Pt: 27 °C
MDL Number: MFCD00051541
CAS Number: 5314-55-6
EINECS: 226-172-9
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty