Ethyl benzilate ≥97.0%
Supplier: TCI America
E0747-5G
TCE0747-5GEA
325
USD
TCE0747-5G
Ethyl benzilate ≥97.0%
Ethyl benzilate
CAS Number: 52182-15-7
MDL Number: MFCD00016850
Molecular Formula: C16H16O3
Molecular Weight: 256.30
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 190
Melting point (°C): 34
Other Product Names / Synonyms: Benzilic Acid Ethyl Ester, Diphenylglycolic Acid Ethyl Ester
Formula:
C₁₆H₁₆O₃ MW: 256.3 g/mol Boiling Pt: 190 °C Melting Pt: 34 °C |
MDL Number:
MFCD00016850 CAS Number: 52182-15-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...