2-Ethoxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
E0723-5G
E0723-1G
TCE0723-5GEA
174.58
USD
TCE0723-5G
TCE0723-1G
2-Ethoxyphenylboronic acid (contains varying amounts of Anhydride)
2-Ethoxyphenylboronic acid
CAS Number: 213211-69-9
MDL Number: MFCD00674027
Molecular Formula: C8H11BO3
Molecular Weight: 165.98
Form: Crystal
Color: White
Melting point (°C): 103
Other Product Names / Synonyms: 2-Ethoxybenzeneboronic Acid
Formula:
C₈H₁₁BO₃ MW: 165.98 g/mol Melting Pt: 103 °C |
MDL Number:
MFCD00674027 CAS Number: 213211-69-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...