TCIAE0687

E0687-5G
TCE0687-5GEA 83.49 USD
TCE0687-5G
TCIAE0687
CAS Number: 64920-29-2
Molecular Formula: C12H14O3
Molecular Weight: 206.24
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 136
Flash Point (°C): 156
Specific Gravity (20/20): 1.09

Other Product Names / Synonyms: Benzylpyruvic Acid Ethyl Ester, Ethyl Benzylpyruvate, 2-Oxo-4-phenylbutyric Acid Ethyl Ester
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR