Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Ethylthiophenol ≥97.0%

Supplier: TCI America


E0672-25G E0672-5G
TCE0672-25GEA 322.52 USD InStock
TCE0672-25G TCE0672-5G
4-Ethylthiophenol ≥97.0%
4-Ethylthiophenol
CAS Number: 4946-13-8
Molecular Formula: C8H10S
Molecular Weight: 138.23
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 211
Specific Gravity (20/20): 1.02

Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents
Other Product Names / Synonyms: 4-Ethylthiophenol


Formula: C₈H₁₀S
MW: 138.23 g/mol
Boiling Pt: 211 °C
Density: 1.02 (20 °C)
MDL Number: MFCD00040229
CAS Number: 4946-13-8
EINECS: 000-000-0
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty