Ethyl-4-Hydroxy-1-piperidinecarboxylate ≥98.0%
Supplier: TCI America
Synonyms:
Ethyl 4-hydroxypiperidine-1-carboxylate
, 1-Carbethoxy-4-hydroxypiperidine
, 4-Hydroxy-1-piperidinecarboxylic Acid Ethyl Ester
E0656-25G
E0656-5G
TCE0656-25GEA
88
USD
TCE0656-25G
TCE0656-5G
Ethyl-4-Hydroxy-1-piperidinecarboxylate ≥98.0%
Ethyl-4-Hydroxy-1-piperidinecarboxylate
CAS Number: 65214-82-6
MDL Number: MFCD00086880
Molecular Formula: C8H15NO3
Molecular Weight: 173.21
Purity/Analysis Method:
Form: Clear Liquid
Specific Gravity (20/20): 1.12
Other Product Names / Synonyms: 1-Carbethoxy-4-hydroxypiperidine, 4-Hydroxy-1-piperidinecarboxylic Acid Ethyl Ester
Formula:
C₈H₁₅NO₃ MW: 173.21 g/mol Density: 1.12 (20 °C) |
MDL Number:
MFCD00086880 CAS Number: 65214-82-6 |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...