Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethoxymethylenemalononitrile ≥98.0%

Supplier: TCI America


E0649-25G
TCE0649-25GEA 54.11 USD InStock
TCE0649-25G
Ethoxymethylenemalononitrile ≥98.0%
Ethoxymethylenemalononitrile
CAS Number: 123-06-8
MDL Number: MFCD00001854
Molecular Formula: C6H6N2O
Molecular Weight: 122.13
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 157
Melting point (°C): 66


Formula: C₆H₆N₂O
MW: 122.13 g/mol
Boiling Pt: 157 °C
Melting Pt: 66 °C
MDL Number: MFCD00001854
CAS Number: 123-06-8
EINECS: 204-597-0
UN: 3439
ADR: 6.1,III
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty