Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

9-Ethynyl-9-fluorenol ≥98.0%

Supplier: TCI America


E0548-5G
TCE0548-5GEA 89.48 USD InStock
TCE0548-5G
9-Ethynyl-9-fluorenol ≥98.0%
9-Ethynyl-9-fluorenol
CAS Number: 13461-74-0
MDL Number: MFCD00041555
Molecular Formula: C15H10O
Molecular Weight: 206.24
Purity/Analysis Method: 98.0% (GC)
Form: Crystal

Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 9-Ethynyl-9-hydroxyfluorene, (9-Hydroxy-9-fluorenyl)acetylene


Formula: C₁₅H₁₀O
MW: 206,25
MDL Number: MFCD00041555
CAS Number: 13461-74-0
EINECS: 000-000-0
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty