Ethyldiphenylphosphine ≥97.0%

Supplier: TCI America

E0519-5G
TCE0519-005GEA 54.81 USD
TCE0519-005G
Ethyldiphenylphosphine ≥97.0%
Ethyldiphenylphosphine

CAS Number: 607-01-2
MDL Number: MFCD00015170
Molecular Formula: C14H15P
Molecular Weight: 214.25
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 157
Flash Point (°C): 110
Specific Gravity (20/20): 1.05

Formula: C₁₄H₁₅P
Boiling Pt: 157 °C
Density: 1.05 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00015170
CAS Number: 607-01-2
UN: 3278
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/80/30/28968030.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR