N-Ethylcyclohexylamine ≥99.0%

Supplier: TCI America

E0506-500ML E0506-25ML
TCE0506-25MLEA 35 USD
TCE0506-25ML TCE0506-500ML
N-Ethylcyclohexylamine ≥99.0%
N-Ethylcyclohexylamine

CAS Number: 5459-93-8
MDL Number: MFCD00003834
Molecular Formula: C8H17N
Molecular Weight: 127.23
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 164
Flash Point (°C): 30
Specific Gravity (20/20): 0.85

Formula: C₈H₁₇N
Boiling Pt: 164 °C
Density: 0.85 (20 °C)
Flash Pt: 30 °C
MDL Number: MFCD00003834
CAS Number: 5459-93-8
https://us.vwr.com/stibo/web/std.lang.all/80/17/28968017.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR