4'-Ethoxyacetophenone ≥98.0%

Supplier: TCI America

E0458-25G
TCE0458-25GEA 133.98 USD
TCE0458-25G
4'-Ethoxyacetophenone ≥98.0%
4'-Ethoxyacetophenone

CAS Number: 1676-63-7
MDL Number: MFCD00009095
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 269
Melting point (°C): 36
Flash Point (°C): 110
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 4-Acetylphenetole

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 269 °C
Melting Pt: 36 °C
Flash Pt: 110 °C
MDL Number: MFCD00009095
CAS Number: 1676-63-7
https://us.vwr.com/stibo/web/std.lang.all/79/73/28967973.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR