Esculetin ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
6,7-Dihydroxycoumarin
E0386-1G
TCE0386-001GEA
47.71
USD
TCE0386-001G
Esculetin ≥98.0% (by HPLC, titration analysis)
Esculetin
CAS Number: 305-01-1
MDL Number: MFCD00006874
Molecular Formula: C9H6O4
Molecular Weight: 178.14
Purity/Analysis Method:
Form: Crystal
Product Categories: Bioscience, Phenylpropanoids, Phenylpropanoids (Phytochemicals), Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: 6,7-Dihydroxycoumarin
Formula:
C₉H₆O₄ MW: 178.14 g/mol |
MDL Number:
MFCD00006874 CAS Number: 305-01-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...