3-Ethoxypropylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

E0296-500ML E0296-25ML
TCE0296-25MLEA 25.38 USD
TCE0296-25ML TCE0296-500ML
3-Ethoxypropylamine ≥98.0% (by GC, titration analysis)
3-Ethoxypropylamine

CAS Number: 6291-85-6
MDL Number: MFCD00008221
Molecular Formula: C5H13NO
Molecular Weight: 103.17
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 40
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: 3-Aminopropyl Ethyl Ether

Formula: C₅H₁₃NO
MW: 103.16 g/mol
Boiling Pt: 132 °C
Density: 0.86 (20 °C)
Flash Pt: 40 °C
MDL Number: MFCD00008221
CAS Number: 6291-85-6
https://us.vwr.com/stibo/web/std.lang.all/78/34/28967834.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR