3-Ethoxysalicylaldehyde ≥97.0%

Supplier: TCI America

E0253-25G
TCE0253-025GEA 18.25 USD
TCE0253-025G
3-Ethoxysalicylaldehyde ≥97.0%
3-Ethoxysalicylaldehyde

CAS Number: 492-88-6
MDL Number: MFCD00003323
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Boiling point (°C): 264
Melting point (°C): 66


Other Product Names / Synonyms: 3-Ethoxy-2-hydroxybenzaldehyde

Formula: C₉H₁₀O₃
Boiling Pt: 264 °C
Melting Pt: 66 °C
MDL Number: MFCD00003323
CAS Number: 492-88-6
https://us.vwr.com/stibo/web/std.lang.all/78/07/28967807.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR