9-Ethylcarbazole ≥99.0%

Supplier: TCI America

Synonyms: 9-Ethyl-9H-carbazoleN-Ethylcarbazole

E0247-25G
TCE0247-025GEA 27 USD
TCE0247-025G
9-Ethylcarbazole ≥99.0%
9-Ethylcarbazole

CAS Number: 86-28-2
MDL Number: MFCD00004967
Molecular Formula: C14H13N
Molecular Weight: 195.27
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 166
Melting point (°C): 68

Formula: C₁₄H₁₃N
MW: 195.27 g/mol
Boiling Pt: 166 °C
Melting Pt: 68 °C
MDL Number: MFCD00004967
CAS Number: 86-28-2
https://us.vwr.com/stibo/web/std.lang.all/78/02/28967802.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR