5-Ethylidene-2-norbornene ≥98.0%

Supplier: TCI America

Synonyms: 5-Ethylidenebicyclo[2.2.1]hept-2-ene

E0244-500ML E0244-25ML
TCE0244-025MLEA 22.31 USD
TCE0244-025ML TCE0244-500ML
5-Ethylidene-2-norbornene ≥98.0%
5-Ethylidene-2-norbornene

CAS Number: 16219-75-3
MDL Number: MFCD00167576
Molecular Formula: C9H12
Molecular Weight: 120.20
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 147
Flash Point (°C): 38
Specific Gravity (20/20): 0.90

Formula: C₉H₁₂
MW: 120.20 g/mol
Boiling Pt: 147 °C
Density: 0.90 (20 °C)
Flash Pt: 38 °C
MDL Number: MFCD00167576
CAS Number: 16219-75-3
https://us.vwr.com/stibo/web/std.lang.all/78/00/28967800.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR