Quinoline ethiodide ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1-Ethylquinolinium Iodide

E0173-25G
TCE0173-025GEA 53.79 USD
TCE0173-025G
Quinoline ethiodide ≥99.0% (by titrimetric analysis)
Quinoline ethiodide

CAS Number: 634-35-5
MDL Number: MFCD00041996
Molecular Formula: C11H12IN
Molecular Weight: 285.13
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Melting point (°C): 157


Other Product Names / Synonyms: 1-Ethylquinolinium Iodide

Formula: C₁₁H₁₂IN
MW: 285.13 g/mol
Melting Pt: 157 °C
MDL Number: MFCD00041996
CAS Number: 634-35-5
https://us.vwr.com/stibo/web/std.lang.all/77/48/28967748.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR