Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethyl-1,4-phenylenediammonium dichloride ≥99.0% (by HPLC, titration analysis), for Biochemical research

Supplier: TCI America

Synonyms: N,N-Dimethyl-p-phenylenediamine dihydrochloride

D3931-5G D3931-1G
TCD3931-5GEA 55.37 USD InStock
TCD3931-5G TCD3931-1G
N,N-Dimethyl-1,4-phenylenediammonium dichloride ≥99.0% (by HPLC, titration analysis), for Biochemical research
N,N-Dimethyl-1,4-phenylenediammonium dichloride
[for Biochemical Research]
CAS Number: 536-46-9
MDL Number: MFCD00012991
Molecular Formula: C8H12N2
Molecular Weight: 209.11
Purity/Analysis Method: 99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 208
Storage Temperature: 0-10°C

Product Categories: Bioscience, Peroxidase Substrates
Other Product Names / Synonyms: 4-Dimethylaminoaniline Dihydrochloride, N,N-Dimethyl-p-phenylenediammonium Dichloride


Formula: (CH₃)₂NC₆H₄NH₂·2HCl
MW: 209,12 g/mol
Melting Pt: 208 °C
Storage Temperature: ambient
MDL Number: MFCD00012991
CAS Number: 536-46-9
EINECS: 208-635-7
UN: 2811
ADR: 6.1,II
Merck Index: 13,03280
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (HPLC,T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty