Diethylzinc ∼15% ∼1 mol/L in toluene
Supplier: TCI America
D3902-100ML
TCD3902-100MLEA
116.41
USD
TCD3902-100ML
Diethylzinc ∼15% ∼1 mol/L in toluene
Diethylzinc
CAS Number: 557-20-0
MDL Number: MFCD00009021
Molecular Formula: C4H10Zn
Molecular Weight: 123.50
Form: Clear Liquid
Color: Colorless
Flash Point (°C): -12
Specific Gravity (20/20): 0.90
Storage Temperature: 0-10°C
Formula:
C₄H₁₀Zn Density: 0.90 (20 °C) Flash Pt: –12 °C |
MDL Number:
MFCD00009021 CAS Number: 557-20-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...