Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(2,3-Difluorophenyl)acetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America


D3889-1G D3889-5G
TCD3889-1GEA 146.69 USD InStock
TCD3889-1G TCD3889-5G
(2,3-Difluorophenyl)acetic acid ≥98.0% (by GC, titration analysis)
(2,3-Difluorophenyl)acetic acid
CAS Number: 145689-41-4
MDL Number: MFCD00040968
Molecular Formula: C8H6F2O2
Molecular Weight: 172.13
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 118


Formula: (C₆H₃F₂)CH₂COOH
MW: 172,13 g/mol
Melting Pt: 118 °C
MDL Number: MFCD00040968
CAS Number: 145689-41-4
EINECS: 000-000-0
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty