N,N-Dimethylaniline ≥99.0% for biochemical research

Supplier: TCI America

D3866-1G D3866-5G
TCD3866-5GEA 36.45 USD
TCD3866-5G TCD3866-1G
N,N-Dimethylaniline ≥99.0% for biochemical research
N,N-Dimethylaniline

[for Biochemical Research]
CAS Number: 121-69-7
MDL Number: MFCD00008304
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 194
Flash Point (°C): 73
Freezing point (°C): 2
Specific Gravity (20/20): 0.96


Product Categories: Bioscience, Peroxidase Substrates
Other Product Names / Synonyms: DMA

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 194 °C
Density: 0.96 (20 °C)
Flash Pt: 73 °C
MDL Number: MFCD00008304
CAS Number: 121-69-7
UN: 2253
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/58/16/28985816.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR