Dodecyl methyl sulfide ≥98.0%

Supplier: TCI America

D3767-5G D3767-25G
TCD3767-25GEA 94.6 USD
TCD3767-25G TCD3767-5G
Dodecyl methyl sulfide ≥98.0%
Dodecyl methyl sulphide

CAS Number: 3698-89-3
MDL Number: MFCD00015073
Molecular Formula: C13H28S
Molecular Weight: 216.43
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Melting point (°C): -2
Flash Point (°C): 110
Specific Gravity (20/20): 0.85

https://us.vwr.com/stibo/web/std.lang.all/57/37/28985737.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR