3,3-Dimethyl-2-oxobutyric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Trimethylpyruvic Acid3,3-Dimethyl-2-oxobutanoic acid

D3609-25G D3609-5G
TCD3609-25GEA 189.81 USD
TCD3609-25G TCD3609-5G
3,3-Dimethyl-2-oxobutyric acid ≥98.0% (by titrimetric analysis)
3,3-Dimethyl-2-oxobutyric acid

CAS Number: 815-17-8
MDL Number: MFCD00154352
Molecular Formula: C6H10O3
Molecular Weight: 130.14
Purity/Analysis Method: 98.0% (T)
Form: Clear Liquid
Boiling point (°C): 80
Freezing point (°C): 13
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: Trimethylpyruvic Acid

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 80 °C
Density: 1.06 (20 °C)
MDL Number: MFCD00154352
CAS Number: 815-17-8
https://us.vwr.com/stibo/web/std.lang.all/55/95/28985595.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR