Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4-Difluoro-2-iodobenzene ≥98.0%

Supplier: TCI America

Synonyms: 2,5-Difluoroiodobenzene

D3594-1G D3594-5G
TCD3594-1GEA 30.17 USD InStock
TCD3594-1G TCD3594-5G
1,4-Difluoro-2-iodobenzene ≥98.0%
1,4-Difluoro-2-iodobenzene
CAS Number: 2265-92-1
MDL Number: MFCD03094151
Molecular Formula: C6H3F2I
Molecular Weight: 239.99
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 183
Flash Point (°C): 85
Specific Gravity (20/20): 2.01


Formula: C₆H₃F₂I
MW: 239,99 g/mol
Boiling Pt: 183 °C
Density: 2,01 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD03094151
CAS Number: 2265-92-1
EINECS: 000-000-0
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty