1,3-Dibromo-5-fluoro-2-iodobenzene ≥98.0%
Supplier: TCI America
D3582-5G
TCD3582-5GEA
77
USD
TCD3582-5G
1,3-Dibromo-5-fluoro-2-iodobenzene ≥98.0%
1,3-Dibromo-5-fluoro-2-iodobenzene
CAS Number: 62720-29-0
MDL Number: MFCD00042232
Molecular Formula: C6H2Br2FI
Molecular Weight: 379.79
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 134
Formula:
C₆H₂Br₂FI MW: 379.79 g/mol Melting Pt: 134 °C |
MDL Number:
MFCD00042232 CAS Number: 62720-29-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...