2',4'-Difluoroacetanilide ≥98.0%
Supplier: TCI America
D3578-25G
TCD3578-25GEA
29.42
USD
TCD3578-25G
2',4'-Difluoroacetanilide ≥98.0%
2',4'-Difluoroacetanilide
CAS Number: 399-36-0
MDL Number: MFCD00032502
Molecular Formula: C8H7F2NO
Molecular Weight: 171.15
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 122
Formula:
C₈H₇F₂NO Melting Pt: 122 °C |
MDL Number:
MFCD00032502 CAS Number: 399-36-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...