Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Dichloro-4-fluorobenzene ≥98.0%

Supplier: TCI America

Synonyms: 3,4-Dichlorofluorobenzene

D3570-25G
TCD3570-25GEA 49.93 USD InStock
TCD3570-25G
1,2-Dichloro-4-fluorobenzene ≥98.0%
1,2-Dichloro-4-fluorobenzene
CAS Number: 1435-49-0
MDL Number: MFCD00018119
Molecular Formula: C6H3Cl2F
Molecular Weight: 164.99
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 168
Melting point (°C): -1
Flash Point (°C): 64
Specific Gravity (20/20): 1.41


Formula: C₆H₃Cl₂F
MW: 164,99 g/mol
Boiling Pt: 168 °C
Melting Pt: -1 °C
Density: 1,41 (20 °C)
Flash Pt: 64 °C
MDL Number: MFCD00018119
CAS Number: 1435-49-0
EINECS: 215-858-3
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty