cis-Decahydro-1H,6H-3a,5a,8a,10a-tetraazapyrene ≥98.0%
Supplier: TCI America
Synonyms:
cis-1,4,8,11-Perhydrotetraazapyrene
D3561-100MG
TCD3561-100MGEA
103.48
USD
TCD3561-100MG
cis-Decahydro-1H,6H-3a,5a,8a,10a-tetraazapyrene ≥98.0%
cis-Decahydro-1H,6H-3a,5a,8a,10a-tetraazapyrene
CAS Number: 74199-16-9
MDL Number: MFCD04038934
Molecular Formula: C12H22N4
Molecular Weight: 222.34
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 83
Formula:
C₁₂H₂₂N₄ MW: 222.34 g/mol Melting Pt: 83 °C |
MDL Number:
MFCD04038934 CAS Number: 74199-16-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...