3,4-Dichloro-1,2,5-thiadiazole ≥98.0%
Supplier: TCI America
D3547-25G
TCD3547-25GEA
58.87
USD
TCD3547-25G
3,4-Dichloro-1,2,5-thiadiazole ≥98.0%
3,4-Dichloro-1,2,5-thiadiazole
CAS Number: 5728-20-1
MDL Number: MFCD00010072
Molecular Formula: C2Cl2N2S
Molecular Weight: 155.00
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 158
Flash Point (°C): 100
Specific Gravity (20/20): 1.65
Formula:
C₂Cl₂N₂S Boiling Pt: 158 °C Density: 1.65 (20 °C) Flash Pt: 100 °C |
MDL Number:
MFCD00010072 CAS Number: 5728-20-1 UN: 2810 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...