6,7-Dihydro-5H-cyclopenta[b]pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
5H-1-Pyrindine, 6,7-dihydro-
, 2,3-Cyclopentenopyridine
D3536-5G
D3536-25G
TCD3536-5GEA
32.48
USD
TCD3536-5G
TCD3536-25G
6,7-Dihydro-5H-cyclopenta[b]pyridine ≥98.0% (by GC, titration analysis)
6,7-Dihydro-5H-cyclopenta[b]pyridine
CAS Number: 533-37-9
MDL Number: MFCD00005933
Molecular Formula: C8H9N
Molecular Weight: 119.17
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 88
Flash Point (°C): 67
Specific Gravity (20/20): 1.04
Other Product Names / Synonyms: 2,3-Cyclopentenopyridine
Formula:
C₈H₉N MW: 119.17 g/mol Boiling Pt: 88 °C Density: 1.04 (20 °C) Flash Pt: 67 °C |
MDL Number:
MFCD00005933 CAS Number: 533-37-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...