2,3-Dimethylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
D3516-5G
TCD3516-5GEA
69.02
USD
TCD3516-5G
2,3-Dimethylphenylboronic acid (contains varying amounts of Anhydride)
2,3-Dimethylphenylboronic acid
CAS Number: 183158-34-1
MDL Number: MFCD01863524
Molecular Formula: C8H11BO2
Molecular Weight: 149.98
Form: Crystal
Color: White
Melting point (°C): 175
Other Product Names / Synonyms: 2,3-Dimethylbenzeneboronic Acid, o-Xylene-3-boronic Acid
Formula:
C₈H₁₁BO₂ MW: 149.98 g/mol Melting Pt: 175 °C |
MDL Number:
MFCD01863524 CAS Number: 183158-34-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...