2,6-Dibromo-p-cresol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3,5-Dibromo-4-hydroxytoluene
, 1,3-Dibromo-2-hydroxy-5-methylbenzene
, 2,6-Dibromo-4-methylphenol
D3487-25G
TCD3487-25GEA
198.94
USD
TCD3487-25G
2,6-Dibromo-p-cresol ≥98.0% (by GC, titration analysis)
2,6-Dibromo-p-cresol
CAS Number: 2432-14-6
MDL Number: MFCD00002154
Molecular Formula: C7H6Br2O
Molecular Weight: 265.93
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 140
Melting point (°C): 48
Flash Point (°C): 113
Other Product Names / Synonyms: 2,6-Dibromo-4-methylphenol
Formula:
C₇H₆Br₂O MW: 265.93 g/mol Boiling Pt: 140 °C Melting Pt: 48 °C Flash Pt: 113 °C |
MDL Number:
MFCD00002154 CAS Number: 2432-14-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...