Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Dimethoxythiophenol ≥93.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1,2-Dimethoxy-4-mercaptobenzene3,4-Dimethoxybenzene-1-thiol4-Mercaptoveratrole3,4-Dimethoxybenzenethiol3,4-Dimethoxyphenyl mercaptan

D3469-5G D3469-25G
TCD3469-5GEA 93.64 USD InStock
TCD3469-5G TCD3469-25G
3,4-Dimethoxythiophenol ≥93.0% (by titrimetric analysis)
3,4-Dimethoxythiophenol
CAS Number: 700-96-9
MDL Number: MFCD00052823
Molecular Formula: C8H10O2S
Molecular Weight: 170.23
Purity/Analysis Method: 93.0% (T)
Form: Clear Liquid
Boiling point (°C): 138
Flash Point (°C): 113
Specific Gravity (20/20): 1.20

Other Product Names / Synonyms: 3,4-Dimethoxythiophenol, 4-Mercaptoveratrole


Formula: C₈H₁₀O₂S
MW: 170,23 g/mol
Boiling Pt: 138 °C
Density: 1.20 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00052823
CAS Number: 700-96-9
EINECS: 000-000-0
UN: 3334
ADR: 9,
Order Now
Specification Test Results
Purity/Analysis Method 93.0% (T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty