2,4-Difluorophenyl isothiocyanate ≥98.0%
Supplier: TCI America
D3454-1G
TCD3454-1GEA
86
USD
TCD3454-1G
2,4-Difluorophenyl isothiocyanate ≥98.0%
2,4-Difluorophenyl isothiocyanate
CAS Number: 141106-52-7
MDL Number: MFCD00041044
Molecular Formula: C7H3F2NS
Molecular Weight: 171.16
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 86
Flash Point (°C): 93
Specific Gravity (20/20): 1.34
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Isothiocyanic Acid 2,4-Difluorophenyl Ester
Formula:
C₇H₃F₂NS MW: 171.17 g/mol Boiling Pt: 86 °C Density: 1.34 (20 °C) Flash Pt: 93 °C |
MDL Number:
MFCD00041044 CAS Number: 141106-52-7 UN: 3334 ADR: 9, |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...