3,4-Dimethylthiophenol ≥97.0% (by GC, titration analysis)
D3344-5G
TCD3344-5GEA
122.1
USD
TCD3344-5G
3,4-Dimethylthiophenol ≥97.0% (by GC, titration analysis)
3,4-Dimethylthiophenol
CAS Number: 18800-53-8
Molecular Formula: C8H10S
Molecular Weight: 138.23
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 218
Flash Point (°C): 90
Specific Gravity (20/20): 1.04
Other Product Names / Synonyms: 3,4-Dimethylthiophenol, 4-Mercapto-o-xylene, o-Xylene-4-thiol
Formula:
C₈H₁₀S MW: 138.23 g/mol Boiling Pt: 218 °C Density: 1.04 (20 °C) Flash Pt: 90 °C |
MDL Number:
MFCD00010023 CAS Number: 18800-53-8 UN: 2810 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...