2,5-Diacetoxytoluene ≥97.0%

Supplier: TCI America

Synonyms: Methylhydroquinone diacetate1,4-Diacetoxy-2-methylbenzene

D3318-25G
TCD3318-25GEA 63 USD
TCD3318-25G
2,5-Diacetoxytoluene ≥97.0%
2,5-Diacetoxytoluene

CAS Number: 717-27-1
Molecular Formula: C11H12O4
Molecular Weight: 208.21
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 49
Flash Point (°C): 110


Other Product Names / Synonyms: 1,4-Diacetoxy-2-methylbenzene, Methylhydroquinone Diacetate

Formula: C₁₁H₁₂O₄
Melting Pt: 49 °C
Flash Pt: 110 °C
MDL Number: MFCD00061098
CAS Number: 717-27-1
https://us.vwr.com/stibo/web/std.lang.all/02/92/28980292.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR