TCIAD3310

D3310-25G D3310-5G
TCD3310-25GEA 456.96 USD
TCD3310-25G TCD3310-5G
TCIAD3310
CAS Number: 26878-35-3
Molecular Formula: C5H7N3
Molecular Weight: 182.05
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Melting point (°C): 246
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR