TCIAD3308

D3308-1G D3308-5G
TCD3308-1GEA 53.09 USD
TCD3308-1G TCD3308-5G
TCIAD3308
CAS Number: 56578-18-8
Molecular Formula: C10H20O
Molecular Weight: 156.27
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 86
Flash Point (°C): 62
Specific Gravity (20/20): 0.85
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR