3,4-Dimethylbenzophenone ≥98.0%

Supplier: TCI America

Synonyms: (3,4-Dimethylphenyl)(phenyl)methanone

D3289-25G
TCD3289-25GEA 48.01 USD
TCD3289-25G
3,4-Dimethylbenzophenone ≥98.0%
3,4-Dimethylbenzophenone

CAS Number: 2571-39-3
Molecular Formula: C15H14O
Molecular Weight: 210.28
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 335
Melting point (°C): 45
Flash Point (°C): 110

Formula: C₁₅H₁₄O
MW: 210.28 g/mol
Boiling Pt: 335 °C
Melting Pt: 45 °C
Flash Pt: 110 °C
MDL Number: MFCD00008525
CAS Number: 2571-39-3
https://us.vwr.com/stibo/web/std.lang.all/02/64/28980264.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR