1,4-Bis(methylamino)-9,10-anthraquinone
Supplier: TCI America
Synonyms:
Disperse Blue 14
, 1,4-Bis(methylamino)anthraquinone
, 1,4-Bis(methylamino)anthracene-9,10-dione
D3280-25G
TCD3280-25GEA
116.94
USD
TCD3280-25G
1,4-Bis(methylamino)-9,10-anthraquinone
1,4-Bis(methylamino)-9,10-anthraquinone
CAS Number: 2475-44-7
MDL Number: MFCD00001198
Molecular Formula: C16H14N2O2
Molecular Weight: 266.30
Form: Crystal
Color: Deep Purple
Melting point (°C): 222
Lambda max.: 643 nm (EtOH)
Other Product Names / Synonyms: 1,4-Bis(methylamino)anthraquinone
Formula:
C₁₆H₁₄N₂O₂ MW: 266.29 g/mol Melting Pt: 222 °C |
MDL Number:
MFCD00001198 CAS Number: 2475-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...