3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Diethyl 3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl PhosphateDEPBT

D3262-5G
TCD3262-5GEA 63.73 USD
TCD3262-5G
3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one ≥98.0% (by HPLC, total nitrogen)
3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one

CAS Number: 165534-43-0
Molecular Formula: C11H14N3O5P
Molecular Weight: 299.22
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 73


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis, Peptide Synthesis
Other Product Names / Synonyms: DEPBT, Diethyl 3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl Phosphate

Formula: C₁₁H₁₄N₃O₅P
Melting Pt: 73 °C
MDL Number: MFCD01236967
CAS Number: 165534-43-0
https://us.vwr.com/stibo/web/std.lang.all/02/47/28980247.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR