3,5-Dichlorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
D2909-5G
TCD2909-5GEA
68
USD
TCD2909-5G
3,5-Dichlorophenylboronic acid (contains varying amounts of Anhydride)
3,5-Dichlorophenylboronic acid
CAS Number: 67492-50-6
MDL Number: MFCD00051935
Molecular Formula: C6H5BCl2O2
Molecular Weight: 190.81
Form: Crystal
Color: White
Melting point (°C): 306
Other Product Names / Synonyms: 3,5-Dichlorobenzeneboronic Acid
Formula:
C₆H₅BCl₂O₂ MW: 190.82 g/mol Melting Pt: 306 °C |
MDL Number:
MFCD00051935 CAS Number: 67492-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...