2,4-Dichlorobenzamide ≥98.0%

Supplier: TCI America

D2867-25G
TCD2867-25GEA 110.64 USD
TCD2867-25G
2,4-Dichlorobenzamide ≥98.0%
2,4-Dichlorobenzamide

CAS Number: 2447-79-2
MDL Number: MFCD00007974
Molecular Formula: C7H5Cl2NO
Molecular Weight: 190.02
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 193

Formula: C₇H₅Cl₂NO
Melting Pt: 193 °C
MDL Number: MFCD00007974
CAS Number: 2447-79-2
https://us.vwr.com/stibo/web/std.lang.all/00/22/28980022.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR