N,N'-(1,2-Dihydroxyethylene)bisacrylamide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1,2-Bisacrylamido-1,2-ethanediolN,N'-Diacryloyl-1,2-dihydroxyethylenediamine

D2864-5G D2864-25G
TCD2864-25GEA 348.84 USD
TCD2864-25G TCD2864-5G
N,N'-(1,2-Dihydroxyethylene)bisacrylamide ≥98.0% (by titrimetric analysis)
N,N'-(1,2-Dihydroxyethylene)bisacrylamide

CAS Number: 868-63-3
MDL Number: MFCD00008624
Molecular Formula: C8H12N2O4
Molecular Weight: 200.19
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Boiling point (°C): 156


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,2-Bisacrylamido-1,2-ethanediol, N,N'-Diacryloyl-1,2-dihydroxyethylenediamine

Formula: C₈H₁₂N₂O₄
Boiling Pt: 156 °C
MDL Number: MFCD00008624
CAS Number: 868-63-3
https://us.vwr.com/stibo/web/std.lang.all/00/19/28980019.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR