3,5-Dimethyl-1-phenyl-1H-pyrazole ≥98.0%

Supplier: TCI America

Synonyms: 3,5-Dimethyl-1-phenylpyrazole3,5-Dimethyl-1-phenyl-1H-pyrazole

D2834-25G
TCD2834-25GEA 343 USD
TCD2834-25G
3,5-Dimethyl-1-phenyl-1H-pyrazole ≥98.0%
3,5-Dimethyl-1-phenyl-1H-pyrazole

CAS Number: 1131-16-4
MDL Number: MFCD00020728
Molecular Formula: C11H12N2
Molecular Weight: 172.23
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.06

Formula: C₁₁H₁₂N₂
MW: 172.23 g/mol
Density: 1,06 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00020728
CAS Number: 1131-16-4

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR