Diphenylsilane ≥98.0%
Supplier: TCI America
D2820-5G
D2820-25G
TCD2820-25GEA
232.44
USD
TCD2820-25G
TCD2820-5G
Diphenylsilane ≥98.0%
Diphenylsilane
CAS Number: 775-12-2
MDL Number: MFCD00003002
Molecular Formula: C12H12Si
Molecular Weight: 184.31
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 97
Flash Point (°C): 98
Specific Gravity (20/20): 1.00
Formula:
C₁₂H₁₂Si MW: 184.31 g/mol Boiling Pt: 97 °C Density: 1.00 (20 °C) Flash Pt: 98 °C |
MDL Number:
MFCD00003002 CAS Number: 775-12-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...