2,5-Difluorobenzyl alcohol ≥98.0%

Supplier: TCI America

Synonyms: (2,5-Difluorophenyl)methanol

D2818-5G
TCD2818-5GEA 122 USD
TCD2818-5G
2,5-Difluorobenzyl alcohol ≥98.0%
2,5-Difluorobenzyl alcohol

CAS Number: 75853-20-2
MDL Number: MFCD00010627
Molecular Formula: C7H6F2O
Molecular Weight: 144.12
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 93
Specific Gravity (20/20): 1.30

Formula: C₇H₆F₂O
MW: 144.12 g/mol
Density: 1.30 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00010627
CAS Number: 75853-20-2
https://us.vwr.com/stibo/web/std.lang.all/99/80/28979980.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR